Mometasone furoate
Mometasone furoate
Mometasone furoate
Thermo Scientific Chemicals
Mometasone furoate
Have Questions?
Catalog NumberQuantity
ACR4621300101 g
Catalog number ACR462130010
Price (MYR)
1,461.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,461.00
EA
Add to cart
Specifications
Chemical Name or MaterialMometasone furoate
CAS83919-23-7
Health Hazard 1GHS Signal Word: Danger
Melting Point218°C to 220°C
Molecular FormulaC27H30Cl2O6