Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gliclazide, Thermo Scientific Chemicals
CAS: 21187-98-4 | C15H21N3O3S | 323.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR461070010
Price (MYR)
585.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
585.00
Add to cart
Chemical Identifiers
CAS14381-51-2
IUPAC Name3-bromobenzene-1,2-diol
Molecular FormulaC6H5BrO2
InChI KeyJPBDMIWPTFDFEU-UHFFFAOYSA-N
SMILESOC1=CC=CC(Br)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
Assay (GC)≥94.0%
Melting Point (clear melt)36-45?C
Water Content (Karl Fischer Titration)<2.0%
FormCrystalline or lumpy powder or crystals