Chlorhexidine, 98%, Thermo Scientific Chemicals
Chlorhexidine, 98%, Thermo Scientific Chemicals
Chlorhexidine, 98%, Thermo Scientific Chemicals
Chlorhexidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorhexidine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR461020050
Price (MYR)
1,963.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,963.00
Add to cart
Specifications
Chemical Name or MaterialChlorhexidine
Melting Point134.0°C to 136.0°C
CAS55-56-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Causes serious eye damage. Very toxic to aquatic life with long lasting effects.