Chlorthalidone, 98%, Thermo Scientific Chemicals
Chlorthalidone, 98%, Thermo Scientific Chemicals
Chlorthalidone, 98%, Thermo Scientific Chemicals
Chlorthalidone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorthalidone, 98%, Thermo Scientific Chemicals
CAS: 77-36-1 | C14H11ClN2O4S | 338.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR460930010
Price (MYR)
1,049.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,049.00
Add to cart
Specifications
Chemical Name or MaterialChlorthalidone
CAS77-36-1
EINECS Number201-022-5
Loss on Drying0.5% max. (1 g, 105°C)
Infrared SpectrumAuthentic