Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
Thermo Scientific Chemicals
Copper(II)-ethylenediamine complex, technical, 1M sol. in water, not for viscosity determination
CAS: 14552-35-3 | C4H18CuN4O2 | 217.76 g/mol
Have Questions?
Catalog NumberQuantity
ACR4606800101 L
Catalog number ACR460680010
Price (MYR)
582.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
582.00
Add to cart
Chemical Identifiers
CAS143-10-2
IUPAC Namedecane-1-thiol
Molecular FormulaC10H22S
InChI KeyVTXVGVNLYGSIAR-UHFFFAOYSA-N
SMILESCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
CommentApparent color may differ on changing container
Appearance (Color)Clear colorless to pale blue or pale green
Refractive Index1.4540-1.4590 @ 20?C
FormLiquid
Assay (GC)≥95.0%