Ribavirin, 98%, Thermo Scientific Chemicals
Ribavirin, 98%, Thermo Scientific Chemicals
Ribavirin, 98%, Thermo Scientific Chemicals
Ribavirin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ribavirin, 98%, Thermo Scientific Chemicals
Ribavirin, CAS# 36791-04-5, is an antiviral agent and a guanosine analog, which stops viral RNA synthesis and mRNA capping.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR460480010
Price (MYR)
473.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
473.00
Add to cart
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
FormFoil, 0.74mm (0.029in) thick, annealed.
Assay from Supplier's CofA>99.4% (metals basis)
General Description
  • Ribavirin is a broad-spectrum antiviral nucleoside analog
  • It inhibits viral genome synthesis
Applications
  • Ribavirin is an antiviral compound
  • Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP), which is involved in GTP biosynthesis
RUO – Research Use Only
General References:
  1. Keppeke, G.D.; et al. IMP/GTP balance modulates cytoophidium assembly and IMPDH activity. Cell Div. 2018, 15,13:5.
  2. Li, W.; Shen, F.; Weber, G.; Ribavirin and quercetin synergistically downregulate signal transduction and are cytotoxic in human ovarian carcinoma cells. Oncol Res. 1999, 11(5), 243-7.