Daidzin, 98%, Thermo Scientific Chemicals
Daidzin, 98%, Thermo Scientific Chemicals
Daidzin, 98%, Thermo Scientific Chemicals
Daidzin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Daidzin, 98%, Thermo Scientific Chemicals
CAS: 552-66-9 | C21H20O9 | 416.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Catalog number ACR460421000
Price (MYR)
819.00
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
819.00
Add to cart
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
GC>=98.5 %
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Appearance (Form)Flakes