Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Methyl palmitoleate, 99%, analytical standard for GC
Thermo Scientific Chemicals
Methyl palmitoleate, 99%, analytical standard for GC
CAS: 1120-25-8 | C17H32O2 | 268.44 g/mol
Have Questions?
Catalog NumberQuantity
ACR4604000101 g
Catalog number ACR460400010
Price (MYR)
1,538.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,538.00
Add to cart
Chemical Identifiers
CAS111-87-5
IUPAC Nameoctan-1-ol
Molecular FormulaC8H18O
InChI KeyKBPLFHHGFOOTCA-UHFFFAOYSA-N
SMILESCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Refractive index1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid