Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR460390050
Price (MYR)
969.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
969.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay from Supplier's CofA≥99.985% (metals basis)
Appearance (Color)Gold to gold-brown to brown
Total Metal Impurities≤100ppm