Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl palmitate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-39-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR460390010
Price (MYR)
449.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.