Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-62-9 | C19H36O2 | 296.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR460380050
Price (MYR)
1,147.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,147.00
Add to cart
Specifications
Chemical Name or MaterialMethyl oleate
CAS112-62-9
Melting Point-20.0°C
Density0.8700g/mL
EINECS Number203-992-5