Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl oleate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 112-62-9 | C19H36O2 | 296.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR460380010
Price (MYR)
520.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
520.00
Add to cart
Chemical Identifiers
CAS6600-40-4
IUPAC Name(2S)-2-aminopentanoic acid
Molecular FormulaC5H11NO2
InChI KeySNDPXSYFESPGGJ-BYPYZUCNSA-N
SMILESCCC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Solubility(5 % in water) Clear colorless
Specific optical rotation+22.0° to +26.0° (20°C, 589 nm) (c=2, 5 N HCl)
Appearance (Form)Powder
Appearance (Color)White