Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl myristate, 99%, analytical standard for GC, Thermo Scientific Chemicals
CAS: 124-10-7 | C15H30O2 | 242.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR460360010
Price (MYR)
475.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Chemical Identifiers
CAS50-70-4
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Sulfated ash=<0.1 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Specific optical rotation+4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.)