Succinic acid disodium salt, Thermo Scientific Chemicals
Succinic acid disodium salt, Thermo Scientific Chemicals
Succinic acid disodium salt, Thermo Scientific Chemicals
Succinic acid disodium salt, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Succinic acid disodium salt, Thermo Scientific Chemicals
CAS: 150-90-3 | C4H4Na2O4 | 162.052 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR459631000
Price (MYR)
304.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
304.00
Add to cart
Specifications
Chemical Name or MaterialSuccinic acid disodium salt
CAS150-90-3
Health Hazard 1Warning
Health Hazard 2GHS H StatementCauses skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Health Hazard 3GHS P StatementIF ON SKIN: Wash with plenty of soap and water.IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician if you feel unwell.Wear protective gloves/protective clothing/eye protection/face protection.