Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
CAS: 106266-06-2 | C23H27FN4O2 | 410.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR459390050
Price (MYR)
1,299.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,299.00
Add to cart
Specifications
Chemical Name or MaterialRisperidone
CAS106266-06-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.