Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Risperidone, 99%, Thermo Scientific Chemicals
CAS: 106266-06-2 | C23H27FN4O2 | 410.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR459390010
Price (MYR)
552.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
552.00
Add to cart
Chemical Identifiers
CAS123-42-2
IUPAC Name4-hydroxy-4-methylpentan-2-one
Molecular FormulaC6H12O2
InChI KeySWXVUIWOUIDPGS-UHFFFAOYSA-N
SMILESCC(=O)CC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4220 to 1.4250 (20°C, 589 nm)
Appearance (Form)Liquid