Mycophenolic acid, 98%
Mycophenolic acid, 98%
Mycophenolic acid, 98%
Mycophenolic acid, 98%
Thermo Scientific Chemicals
Mycophenolic acid, 98%
CAS: 24280-93-1 | C17H20O6 | 320.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR459380050
Price (MYR)
909.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
909.00
Add to cart
Chemical Identifiers
CAS104-15-4
IUPAC Name4-methylbenzene-1-sulfonic acid
Molecular FormulaC7H8O3S
InChI KeyJOXIMZWYDAKGHI-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow to pink
Titration with NaOH11 to 13 %