Riluzole, 98%, Thermo Scientific Chemicals
Riluzole, 98%, Thermo Scientific Chemicals
Riluzole, 98%, Thermo Scientific Chemicals
Riluzole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Riluzole, 98%, Thermo Scientific Chemicals
CAS: 1744-22-5 | C8H5F3N2OS | 234.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR459360010
Price (MYR)
854.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
854.00
Add to cart
Chemical Identifiers
CAS16004-15-2
IUPAC Name1-(bromomethyl)-4-iodobenzene
Molecular FormulaC7H6BrI
InChI KeyBQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILESBrCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)75.0-84.0?C
Assay (GC)≥96.0%