βmethasone, 97%
βmethasone, 97%
βmethasone, 97%
Thermo Scientific Chemicals
βmethasone, 97%
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ACR459070250
Price (MYR)
4,304.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
4,304.00
Add to cart
Specifications
Chemical Name or Materialβmethasone
CAS378-44-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May damage the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.