Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR459015000
Price (MYR)
1,046.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,046.00
Add to cart
Chemical Identifiers
CAS531-81-7
IUPAC Name2-oxo-2H-chromene-3-carboxylic acid
Molecular FormulaC10H6O4
InChI KeyACMLKANOGIVEPB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC=CC=C2OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder