Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99%, stabilized, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR459011000
Price (MYR)
446.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
446.00
EA
Add to cart
Specifications
Chemical Name or MaterialIodomethane
CAS74-88-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
CAS7440-50-8