Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Propionyl chloride, 99.5%, phosgene free, Thermo Scientific Chemicals
CAS: 79-03-8 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR458970250
Price (MYR)
368.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
368.00
Add to cart
Specifications
Chemical Name or MaterialPropionyl chloride
CAS79-03-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Reacts violently with water.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call