Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ACR458840250
Price (MYR)
1,565.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,565.00
Add to cart
Chemical Identifiers
CAS60-29-7
IUPAC Nameethoxyethane
Molecular FormulaC4H10O
InChI KeyRTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILESCCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSubmicron filtered [US]
Optical Absorbance at220nm: ≥10% transmittance [UK]
Optical Absorbance at270nm: ≥95% transmittance [UK]
Optical Absorbance at285-400nm: ≥99% transmittance [UK]
Peroxide content≤5 ppm (0.0005%) at time of packaging [US & UK]