Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Simvastatin, 98%, Thermo Scientific Chemicals
CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol
Have Questions?
Catalog NumberQuantity
ACR4588400505 g
Catalog number ACR458840050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 260 nm A: =<0.03
Appearance (Form)Crystalline powder or crystals
Water=<1 %
Arsenic (As)=<5 ppm
Iron (Fe)=<1 ppm