Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ACR458570250
Price (MYR)
785.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
785.00
Add to cart
Chemical Identifiers
CAS94-75-7
IUPAC Name2-(2,4-dichlorophenoxy)acetic acid
Molecular FormulaC8H6Cl2O3
InChI KeyOVSKIKFHRZPJSS-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH96.0 to 104.0 %
Melting point136°C to 141°C
HPLC>=96.0 %