Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylsilanol, Thermo Scientific Chemicals
CAS: 1066-40-6 | C3H9LiOSi | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR458570050
Price (MYR)
472.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
472.00
Add to cart
Chemical Identifiers
CAS146000-39-7
IUPAC Name2-{[(tert-butoxy)carbonyl](methyl)amino}-2-methylpropanoic acid
Molecular FormulaC10H19NO4
InChI KeyKPDLITGXUYMJEC-UHFFFAOYSA-N
SMILESCN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.0 %
Appearance (Color)White
Infrared spectrumConforms
Melting point153°C to 157°C
Appearance (Form)Crystalline powder