Omeprazole, 98+%
Omeprazole, 98+%
Omeprazole, 98+%
Thermo Scientific Chemicals
Omeprazole, 98+%
CAS: 73590-58-6 | C17H19N3O3S | 345.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR458520050
Price (MYR)
521.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
521.00
Add to cart
Chemical Identifiers
CAS565-71-9
IUPAC Name3-amino-2-hydroxypropanoic acid
Molecular FormulaC3H7NO3
InChI KeyBMYNFMYTOJXKLE-UHFFFAOYNA-N
SMILESNCC(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)>97.5%
Elemental Analysis<13.1% Nitrogen
Elemental Analysis<33.71% Carbon
Elemental Analysis<6.79% Hydrogen