Omeprazole, 98+%
Omeprazole, 98+%
Omeprazole, 98+%
Thermo Scientific Chemicals
Omeprazole, 98+%
CAS: 73590-58-6 | C17H19N3O3S | 345.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR458520010
Price (MYR)
437.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
437.00
Add to cart
Specifications
Chemical Name or MaterialOmeprazole
CAS73590-58-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee