Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Donepezil hydrochloride, Thermo Scientific Chemicals
CAS: 120011-70-3 | C24H30ClNO3 | 415.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR458050010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7782-42-5
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.