Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate, 97%
Product Image
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid dipotassium magnesium salt dihydrate, 97%
CAS: 15708-48-2 | C10H12K2MgN2O8 | 390.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR457945000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS104-85-8
IUPAC Name4-methylbenzonitrile
Molecular FormulaC8H7N
InChI KeyVCZNNAKNUVJVGX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystalline mass or fused solid
Assay (GC)≥98.0%