Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™, Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™, Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™, Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Catalog number ACR457070010
Price (MYR)
1,259.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,259.00
Add to cart
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or needles or adhering crystals
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤2%