Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
CAS: 95058-81-4 | C9H11F2N3O4 | 263.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR456890050
Price (MYR)
1,749.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,749.00
Add to cart
Chemical Identifiers
CAS80-09-1
IUPAC Name4-(4-hydroxybenzenesulfonyl)phenol
Molecular FormulaC12H10O4S
InChI KeyVPWNQTHUCYMVMZ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink
Appearance (Form)Powder
Melting point>=245°C
HPLC>=99.6 %
Water=<0.5 % (K.F.)