Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gemcitabine, 98%, Thermo Scientific Chemicals
CAS: 95058-81-4 | C9H11F2N3O4 | 263.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR456890010
Price (MYR)
775.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
775.00
Add to cart
Chemical Identifiers
CAS13453-07-1
IUPAC Namegold hydrochloridyl
Molecular FormulaAuCl
InChI KeyBRSVJNYNWNMJKC-UHFFFAOYSA-N
SMILES[Cl].[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)64.4-65.5% Au