Montelukast sodium, 98%
Montelukast sodium, 98%
Montelukast sodium, 98%
Montelukast sodium, 98%
Thermo Scientific Chemicals
Montelukast sodium, 98%
CAS: 151767-02-1 | C35H35ClNNaO3S | 608.17 g/mol
Have Questions?
Catalog NumberQuantity
ACR4564400101 g
Catalog number ACR456440010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.45-0.55%
Particle Size4-12 mesh
Appearance (Color)Black
Total Metal Impurities≤0.01%
Surface Area900-1100 m²/g