2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™
Acros Organics
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane, 97%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR456400010
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane
CAS83558-87-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF