Cypermethrin, 96%, Thermo Scientific Chemicals
Cypermethrin, 96%, Thermo Scientific Chemicals
Cypermethrin, 96%, Thermo Scientific Chemicals
Cypermethrin, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cypermethrin, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR456311000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCypermethrin
CAS52315-07-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and