Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ACR456290250
Price (MYR)
3,036.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
3,036.00
Add to cart
Chemical Identifiers
CAS25322-68-3
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Hydroxyl Value17-21mg KOH/g
Viscosity200-270 mPas (50%, Water @ 20?C)
Appearance (Color)White to cream
CommentAverage mol weight 5400 - 6600 (based on OH value)
Melting Point (clear melt)55-63?C