Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Forchlorfenuron, 98%, Thermo Scientific Chemicals
CAS: 68157-60-8 | C12H10ClN3O | 247.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ACR456290010
Price (MYR)
555.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
555.00
Add to cart
Specifications
Chemical Name or MaterialForchlorfenuron
CAS68157-60-8
Quantity1 g
Health Hazard 1Warning
Health Hazard 2GHS H Statement Suspected of causing cancer. Toxic to aquatic life with long lasting effects.