Docetaxel, 98%
Docetaxel, 98%
Docetaxel, 98%
Thermo Scientific Chemicals
Docetaxel, 98%
CAS: 114977-28-5 | C43H53NO14 | 807.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number ACR456262500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS533-17-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)84.5-90.5?C
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder