Docetaxel, 98%
Docetaxel, 98%
Docetaxel, 98%
Thermo Scientific Chemicals
Docetaxel, 98%
CAS: 114977-28-5 | C43H53NO14 | 807.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number ACR456260010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS627-63-4
IUPAC Name(2E)-but-2-enedioyl dichloride
Molecular FormulaC4H2Cl2O2
InChI KeyZLYYJUJDFKGVKB-OWOJBTEDSA-N
SMILESClC(=O)C=CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
Refractive Index1.4970-1.5030 @ 20°C