Lutein, 90%
Lutein, 90%
Lutein, 90%
Thermo Scientific Chemicals
Lutein, 90%
CAS: 127-40-2 | C40H56O2 | 568.87 g/mol
Have Questions?
Catalog NumberQuantity
ACR4561600101 g
Catalog number ACR456160010
Price (MYR)
2,116.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
2,116.00
Add to cart
Chemical Identifiers
CAS579-43-1
IUPAC Name1,2-diphenylethane-1,2-diol
Molecular FormulaC14H14O2
InChI KeyIHPDTPWNFBQHEB-UHFFFAOYNA-N
SMILESOC(C(O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and or lumps
Melting point134°C to 140°C
Infrared spectrumConforms
Appearance (Color)White
GC>=94 %