Oxamyl, 95%, Thermo Scientific™
Product Image
Thermo Scientific Chemicals
Oxamyl, 95%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR456140010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialOxamyl
CAS23135-22-0
Quantity1 g
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement Fatal if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.