Oleanolic acid, 97%, Thermo Scientific Chemicals
Oleanolic acid, 97%, Thermo Scientific Chemicals
Oleanolic acid, 97%, Thermo Scientific Chemicals
Oleanolic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Oleanolic acid, 97%, Thermo Scientific Chemicals
CAS: 508-02-1 | C30H48O3 | 456.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR456120050
Price (MYR)
800.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
800.00
Add to cart
Chemical Identifiers
CAS6923-52-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Chloride content< 20 ppm
Elemental Analysis% Sb 40 - 42
Trace Metals< 20 ppm Fe
Formcrystalline powder