1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline-5,6-dione, 98%, Thermo Scientific Chemicals
CAS: 27318-90-7 | C12H6N2O2 | 210.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR456080010
Price (MYR)
1,096.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,096.00
Add to cart
Chemical Identifiers
CAS33693-84-4
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or granules
Appearance (Color)White to cream to yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)81.0-91.0?C
Assay (Silylated GC)≥96.0%