Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Acros Organics
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
1 kg
Catalog number ACR456070050
Price (MYR)
-
Quantity:
5 kg
Request bulk or custom format
Chemical Identifiers
CAS613-92-3
IUPAC Name(Z)-N'-hydroxybenzenecarboximidamide
Molecular FormulaC7H8N2O
InChI KeyMXOQNVMDKHLYCZ-UHFFFAOYSA-N
SMILESN\C(=N/O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Assay (HPLC)≥94.0%
Melting Point (clear melt)70.0-79.0?C