Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Acros Organics
Montmorillonite K-10, granules, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
5 kg
Catalog number ACR456070010
Price (MYR)
162.31
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
162.31
Add to cart
Chemical Identifiers
CAS14024-18-1
IUPAC Nameiron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21FeO6
InChI KeyAQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration Iodometric>=99.0 %
Appearance (Color)Dark orange to dark red