Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ACR455830050
Price (MYR)
1,461.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,461.00
Add to cart
Chemical Identifiers
CAS69-65-8
IUPAC Namehexane-1,2,3,4,5,6-hexol
Molecular FormulaC6H14O6
InChI KeyFBPFZTCFMRRESA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<0.3 % (105°C, 4 hrs)
Specific optical rotation+23° to +25° (20°C, 589 nm) (Ph.Eur.-directives)
HPLC>=98.0 %
Heavy metals (as Pb)=<5 ppm