Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyrimidine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
CAS: 31519-62-7 | C5H4N2O2 | 124.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ACR455830010
Price (MYR)
668.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
668.00
Add to cart
Chemical Identifiers
CAS18618-55-8
IUPAC Nametrichlorocerium heptahydrate
Molecular FormulaCeCl3H14O7
InChI KeyKPZSTOVTJYRDIO-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.O.Cl[Ce](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
CommentMaterial Sourced in UK and US
Identification (FTIR)Conforms (UK Sourced Material)