Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Arbutin, 98%, Thermo Scientific Chemicals
CAS: 497-76-7 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR455640050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7789-78-8
IUPAC Namecalcium
Molecular FormulaCa
InChI KeyOYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Magnesium (Mg)=<8000 ppm
Aluminium (Al)=<100 ppm
Hydrogen (H)>=980 ml/g CaH2
Particle size10 to 100 mm (containing up to 10% fines)