Ketoconazole, 98%
Ketoconazole, 98%
Ketoconazole, 98%
Thermo Scientific Chemicals
Ketoconazole, 98%
Ketoconazole, CAS # 65277-42-1, is a synthetic, broad-spectrum antifungal agent with the ability to inhibit a variety of fungal infections.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ACR455470250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1711-06-4
IUPAC Name3-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyYHOYYHYBFSYOSQ-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.5475-1.5505 @ 20°C
General Description
  • Ketoconazole is an imidazole antifungal agent
  • It functions by inhibiting synthesis of ergosterol, which is the fungal equivalent of cholesterol
Applications
  • Ketoconazole shows fungicide activity against a variety of yeast infections
  • It is an inhibitor of CYP3A4, which is necessary for the conversion of lanosterol to ergosterol
  • It increases fungal cellular permeability
  • It also inhibits endogenous respiration by disrupting membrane phospholipids
  • Ketoconazole can also inhibit the synthesis of thromboxane and sterols such as aldosterone, cortisol, and testosterone
RUO – Research Use Only
General References:
  1. Takano, M.; Hasegawa, R.; Fukuda, T.; Yumoto, R.; Nagai, J.; Murakami, T. Interaction with P-glycoprotein and transport of erythromycin, midazolam and ketoconazole in Caco-2 cells. European Journal of Pharmacology.1998, 358 (3), 289-294.
  2. Goeders, N. E.; Peltier, R. L.; Guerin, G. F. Ketoconazole reduces low dose cocaine self-administration in rats. Drug and Alcohol Dependence.1998, 53 (1), 67-77.